Avantor® Hichrom HI-WAX Plus, GC Columns
Supplier: Avantor
HI14-18-018-10EA
518
EUR
HI14-18-018-10
HI14-18-018-20
HI14-18-018-40
HI14-18-018-50
HI14-18-018-60
HI14-18-020-10
HI14-18-020-20
HI14-18-020-40
HI14-18-020-50
HI14-18-020-60
HI14-18-040-10
HI14-18-040-20
HI14-18-040-40
HI14-18-040-50
HI14-18-040-60
HI14-20-010-10
HI14-20-010-20
HI14-20-010-40
HI14-20-010-50
HI14-20-010-60
HI14-20-020-10
HI14-20-020-20
HI14-20-020-40
HI14-20-020-50
HI14-20-020-60
HI14-20-040-10
HI14-20-040-20
HI14-20-040-40
HI14-20-040-50
HI14-20-040-60
HI14-20-100-10
HI14-20-100-20
HI14-20-100-40
HI14-20-100-50
HI14-20-100-60
HI14-25-010-15
HI14-25-010-30
HI14-25-010-50
HI14-25-010-60
HI14-25-025-10
HI14-25-025-105
HI14-25-025-15
HI14-25-025-30
HI14-25-025-50
HI14-25-025-60
HI14-25-050-10
HI14-25-050-105
HI14-25-050-15
HI14-25-050-30
HI14-25-050-50
HI14-25-050-60
HI14-25-100-10
HI14-25-100-105
HI14-25-100-15
HI14-25-100-30
HI14-25-100-50
HI14-25-100-60
HI14-32-010-10
HI14-32-010-105
HI14-32-010-15
HI14-32-010-30
HI14-32-010-50
HI14-32-010-60
HI14-32-025-10
HI14-32-025-105
HI14-32-025-15
HI14-32-025-30
HI14-32-025-50
HI14-32-025-60
HI14-32-050-10
HI14-32-050-105
HI14-32-050-15
HI14-32-050-30
HI14-32-050-50
HI14-32-050-60
HI14-32-100-10
HI14-32-100-105
HI14-32-100-15
HI14-32-100-30
HI14-32-100-50
HI14-32-100-60
HI14-32-150-10
HI14-32-150-105
HI14-32-150-15
HI14-32-150-30
HI14-32-150-50
HI14-32-150-60
HI14-32-300-10
HI14-32-300-105
HI14-32-300-15
HI14-32-300-30
HI14-32-300-50
HI14-32-300-60
HI14-53-010-10
HI14-53-010-15
HI14-53-010-30
HI14-53-025-15
HI14-53-025-30
HI14-53-025-50
HI14-53-025-60
HI14-53-050-10
HI14-53-050-15
HI14-53-050-30
HI14-53-050-50
HI14-53-050-60
HI14-53-100-10
HI14-53-100-15
HI14-53-100-30
HI14-53-100-50
HI14-53-100-60
HI14-53-100-75
HI14-53-150-10
HI14-53-150-15
HI14-53-150-30
HI14-53-150-50
HI14-53-150-60
HI14-53-150-75
HI14-53-200-10
HI14-53-200-15
HI14-53-200-30
HI14-53-200-50
HI14-53-200-60
HI14-53-200-75
HI14-53-250-10
HI14-53-250-15
HI14-53-250-30
HI14-53-250-50
HI14-53-250-60
HI14-53-250-75
HI14-05-005-2
HI14-05-005-2.5
HI14-05-005-5
HI14-10-010-10
HI14-10-010-15
HI14-10-010-5
HI14-10-020-10
HI14-10-020-15
HI14-10-020-5
HI14-10-040-10
HI14-10-040-15
HI14-10-040-5
HI14-15-015-10
HI14-15-015-15
HI14-15-015-20
HI14-15-015-30
HI14-15-015-5
HI14-25-025-15G5
HI14-25-025-30G5
HI14-25-025-60G5
HI14-53-100-30G5
Avantor® Hichrom HI-WAX Plus, GC Columns
Chromatography Columns
SPECIFICATIONS
Phase name | HI-WAX Plus |
Environmentally Preferable | |
Phase composition | Polyethylene Glycol (PEG) |
Polarity | Polar stationary phase |
Crossbond | Yes |
USP code | G14, G15, G16, G20, G39 |