High visiblity contrast traffic jackets, S426
Supplier: Portwest
S426ONRM S426ONRL S426ONRXL S426ONRXXL S426ONRXXXL S426ONR4XL S426ONRS S426YNR4XL S426YNRL S426YNRM S426YNRS S426YNRXL S426YNRXXL S426YNRXXXL
PRTWS426ONRMEA 155 EUR
PRTWS426ONRM PRTWS426ONRL PRTWS426ONRXL PRTWS426ONRXXL PRTWS426ONRXXXL PRTWS426ONR4XL PRTWS426ONRS PRTWS426YNR4XL PRTWS426YNRL PRTWS426YNRM PRTWS426YNRS PRTWS426YNRXL PRTWS426YNRXXL PRTWS426YNRXXXL
High visiblity contrast traffic jackets, S426
Workwear
A highly innovative and functional jacket that provides seven wearer combinations. This extremely versatile jacket is highly adaptable to all weather conditions and is certified to the highest in waterproof and breathable standards.
  • Breathable garment
  • Taped seams and print access
  • Phone pocket
  • Detachable hood
  • Adaptable in all types of weather
  • Secure,waterproof storage solutions
300D Breathable include fully waterproof taped seams throughout; anti-wicking fabric at hems and cuffs, mobile phone pockets designed to fit a smart phone, good quality zips and strong metal press fasteners; concealed hood under collar; knitted cuffs in sleeves.
Features include fleece lined collar for optimum warmth and comfort, radio loop, D-ring, ID holder, multiple pockets for ample storage, reflective piping, detachable hood for corporate branding
Shell fabric: 100% polyester, 300D oxford weave with a stain resistant finish, PU coated, 190 g
Lining fabric: Outer Jacket: 100% polyester mesh, 60 g; Inner jacket: 65% polyester, 35% cotton, 70 g
Filling fabric: Inner jacket: 100% polyester, 160 g
Certifications: EN ISO 20471 Class 3, EN 343 Class 3:3, GO/RT 3279 Issue 8 (orange only).