High visibility extreme rain trousers, Portwest PWR™, S597
Supplier: Portwest
S597ORRL S597ORRM S597ORRS S597ORRXL S597ORRXXL S597ORRXXXL S597YERL S597YERM S597YERS S597YERXL S597YERXXL S597YERXXXL
PRTWS597ORRLEA 50.7 EUR
PRTWS597ORRL PRTWS597ORRM PRTWS597ORRS PRTWS597ORRXL PRTWS597ORRXXL PRTWS597ORRXXXL PRTWS597YERL PRTWS597YERM PRTWS597YERS PRTWS597YERXL PRTWS597YERXXL PRTWS597YERXXXL
High visibility extreme rain trousers, Portwest PWR™, S597
Workwear
Waterproof, windproof and breathable trousers, features an elasticated waistband and inner drawcord adjustment for a comfortable fit. Undergarments can be easily accessed through side access pockets. Zip fastening at the side leg allows for easy fitting over work boots.
  • Premium stain resistant finish repels oil, water and grime
  • Taped seams to prevent water penetration
  • Reflective tape for increased visibility
  • Side access pockets and easy access cargo pockets
  • Reflective piping for increased visibility
  • Drawcord adjustable waist for a comfortable fit
PWR™ is a premium high performance rainwear fabric that offers superior comfort, function and endurance in all weather conditions.
Weight: 200 g/m²
Certifications: EN ISO 20471 Class 2 and ANSI standards.This collection far exceeds the requirements of EN 343:2019 Class 4:3 R WP 11000 mm MVP 50000 g/m²/24 hrs. RIS-3279-TOM ISSUE 1 (orange only).
Caution: Wash with like colours. Do not dry clean.