Rain jackets, S530, Technik™
Supplier: Portwest
S530NARXXL S530NAR5XL S530NARS S530NARXS S530NARL S530NARM S530NARXL S530NARXXXL S530NAR4XL
PRTWS530NARXXLEA 74.3 EUR
PRTWS530NARXXL PRTWS530NAR5XL PRTWS530NARS PRTWS530NARXS PRTWS530NARL PRTWS530NARM PRTWS530NARXL PRTWS530NARXXXL PRTWS530NAR4XL
Rain jackets, S530, Technik™
Workwear
An excellent jacket for leisure or work. A luxury feel microfibre peach finish is twinned with a 280 g fleece lining that keeps in heat and feels comfortable to wear. Features include double storm flaps, drawstring hood and waist and lower zip pockets.
  • Waterproof keeping the wearer dry and protected from the elements
  • Breathable fabric to draw moisture away from the body keeping the wearer cool, dry and comfortable
  • Four pockets for ample storage and one phone pocket
  • Two-way zip for quick and easy access
  • Pack away hood for added functionality
  • Hook and loop cuffs for a secure fit
Shell fabric: 100% polyester microfibre, peach finish, breathable, PU coated, 140 g
Lining fabric: body: 100% polyester fleece, 280 g; sleeves: 100% polyester, 60 g
Filling fabric: sleeves: 100% polyester filling, 100 g
Certifications: EN 343 Class 3:2