High visibility traffic jackets, RT30
Supplier: Portwest
RT30ORR5XL RT30ORRXS RT30ORRS RT30ORRM RT30ORRL RT30ORRXL RT30ORR4XL RT30ORRXXL RT30ORRXXXL
PRTWRT30ORR5XLEA 51.7 EUR
PRTWRT30ORR5XL PRTWRT30ORRXS PRTWRT30ORRS PRTWRT30ORRM PRTWRT30ORRL PRTWRT30ORRXL PRTWRT30ORR4XL PRTWRT30ORRXXL PRTWRT30ORRXXXL
High visibility traffic jackets, RT30
Workwear
These jackets meets the stringent safety requirements the standard requires.
  • Waterproof with taped seams preventing water penetration
  • Reflective tape for increased visibility
  • Extremely water resistant fabric finish, water beads away from fabric surface
  • Fully lined and padded to trap the heat and increase warmth
  • Four pockets for ample storage
  • Concealed mobile phone pocket
  • Internal chest pocket
  • Two-way zip for quick and easy access
  • Storm flap front to protect against the elements
Shell fabric: 300D Industry 190 g
Lining fabric : 100% Polyester 60 g
Filling fabric : 100% Polyester 170 g
Certifications: CE certified, ANSI/ISEA 107: 2015 TYPE R CLASS 3, EN 343:2019 Class 3:1 X WP 15,000mm, EN 342 0.345 (M².K/W), 2, X, EN ISO 20471 Class 3, RIS-3279-TOM Issue 1.