T-shirts, long sleeves, FR11
Supplier: Portwest
FR11NAR5XL FR11NARM FR11NARXL FR11NARXXL FR11NARXS FR11NARXXXL FR11NAR4XL FR11NARL FR11NARS
PRTWFR11NAR5XLEA 61.8 EUR
PRTWFR11NAR5XL PRTWFR11NARM PRTWFR11NARXL PRTWFR11NARXXL PRTWFR11NARXS PRTWFR11NARXXXL PRTWFR11NAR4XL PRTWFR11NARL PRTWFR11NARS
T-shirts, long sleeves, FR11
Workwear
High quality construction and premium materials set this flame-resistant, anti-static long sleeve T-shirt.
  • Inherent flame resistant qualities will not diminish with washing
  • Flame resistant ribbed cuff for added safety
  • Protection against radiant, convective and contact heat
  • Outstanding colour fastness and shrinkage results
  • Breathable fabric to draw moisture away from the body keeping the wearer cool, dry and comfortable
  • 50+ UPF rated fabric to block 98% of UV rays
  • Ribbed cuffs for warmth and comfort
This industry leading range provides multi standard protection for hazardous environments. These state of the art products are the result of years of experience combined with advanced technology and market research. Commitment to the health, safety and comfort of the wearer can be seen in the wide range of products suitable for all climates and end uses.
Weight: 200 g
Certifications: EN 1149 -5; EN ISO 11612 (A1, B1, C1, F1); IEC 61482-2 IEC 61482-1-2 Class 1; ASTM F1959/F1959M-12 (EBT = 4,3 Cal/cm² (HAF = 66%)).