ESD safety shoes, Slip-On, uvex 1 sport hygiene, S2, 6580
Supplier: UVEX
6580.7/35 6580.7/36 6580.7/37 6580.7/38 6580.7/39 6580.7/40 6580.7/41 6580.7/42 6580.7/43 6580.7/44 6580.7/45 6580.7/46 6580.7/47 6580.7/48 6580.7/49 6580.7/50 6580.7/51 6580.7/52 6580.8/35 6580.8/36 6580.8/37 6580.8/38 6580.8/39 6580.8/40 6580.8/41 6580.8/42 6580.8/43 6580.8/44 6580.8/45 6580.8/46 6580.8/47 6580.8/48 6580.8/49 6580.8/50 6580.8/51 6580.8/52 6580.9/35 6580.9/36 6580.9/37 6580.9/38 6580.9/39 6580.9/40 6580.9/41 6580.9/42 6580.9/43 6580.9/44 6580.9/45 6580.9/46 6580.9/47 6580.9/48 6580.9/49 6580.9/50 6580.9/51 6580.9/52
UVEX6580.7/35EA 93.1 EUR
UVEX6580.7/35 UVEX6580.7/36 UVEX6580.7/37 UVEX6580.7/38 UVEX6580.7/39 UVEX6580.7/40 UVEX6580.7/41 UVEX6580.7/42 UVEX6580.7/43 UVEX6580.7/44 UVEX6580.7/45 UVEX6580.7/46 UVEX6580.7/47 UVEX6580.7/48 UVEX6580.7/49 UVEX6580.7/50 UVEX6580.7/51 UVEX6580.7/52 UVEX6580.8/35 UVEX6580.8/36 UVEX6580.8/37 UVEX6580.8/38 UVEX6580.8/39 UVEX6580.8/40 UVEX6580.8/41 UVEX6580.8/42 UVEX6580.8/43 UVEX6580.8/44 UVEX6580.8/45 UVEX6580.8/46 UVEX6580.8/47 UVEX6580.8/48 UVEX6580.8/49 UVEX6580.8/50 UVEX6580.8/51 UVEX6580.8/52 UVEX6580.9/35 UVEX6580.9/36 UVEX6580.9/37 UVEX6580.9/38 UVEX6580.9/39 UVEX6580.9/40 UVEX6580.9/41 UVEX6580.9/42 UVEX6580.9/43 UVEX6580.9/44 UVEX6580.9/45 UVEX6580.9/46 UVEX6580.9/47 UVEX6580.9/48 UVEX6580.9/49 UVEX6580.9/50 UVEX6580.9/51 UVEX6580.9/52
ESD safety shoes, Slip-On, uvex 1 sport hygiene, S2, 6580
Shoes and Boots
Modern, metal-free, ultra-lightweight and flexible S2 safety slip-on shoe with removable hook-and-loop strap.
  • Upper made from water repellent microfibre material that is easy to clean
  • Minimal number of seams to reduce contamination
  • Suitable for people allergic to chrome as it’s made from synthetic materials
  • Washable at 30 °C
  • All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Soft padding on collar and tongue
100% metal-free uvex xenova® toe cap, with a compact design, anatomical shape, good lateral stability, and no thermal conductivity. The ergonomically designed outsole is comprised of dual-density polyurethane with good shock absorption and excellent slip resistance.
A removable anti-static comfortable insole provides the foot with exceptional, full-surface shock absorption and ensures comfort in high-strain zones.
Certifications: EN ISO 20345:2011 S2 SRC.