VWR® Stainless Steel Erlenmeyer Flask Clamps
Supplier: VWR Collection
444-7040EA 42.2 EUR
444-7040 444-7041 444-7042 444-7043 444-7044 444-7045
VWR® Stainless Steel Erlenmeyer Flask Clamps
Clamps General Purpose Clamps
Accessories information: * Platform supplied with mini shaker and incubating mini shaker. Note other shakers require an optional universal platform to enable flask clamps or test tube racks to be mounted which must be ordered separately.