VWR® Stainless Steel Media Bottle Clamps
Supplier: VWR Collection
444-7035EA 171 EUR
444-7035 444-7036
VWR® Stainless Steel Media Bottle Clamps
Clamps General Purpose Clamps
Accessories information: * Platform supplied with mini shaker and incubating mini shaker. Note other shakers require an optional perforated platform to enable flask clamps or test tube racks to be mounted which must be ordered separately.