Racks for chest freezers, Classic
Supplier: TENAK
TE21440 TE21457 TE21467 TE21470 TE21468 TE21466 TE21465 TE21464 TE21462 TE21474 TE21458 TE21478 TE21456 TE21455 TE21454 TE21452 TE21451 TE21450 TE21449 TE21448 TE21441 TE21460 TE21472 TE21357 TE21356 TE21358 TE21360 TE21362 TE21354 TE21350 TE21352 TE21430 TE21432 TE21466B TE21454B TE21456B TE21472B TE21474B TE21448B TE21440B TE21441B TE21449B TE21450B TE21451B TE21452B TE21455B TE21457B TE21458B TE21460B TE21462B TE21464B TE21467B TE21468B TE21470B TE21478B
479-0644EA 83.4 EUR
479-0644 479-0654 479-0661 479-0663 479-0662 479-0660 479-0659 479-0658 479-0657 479-0665 479-0655 479-0666 479-0653 479-0652 479-0651 479-0650 479-0649 479-0648 479-0647 479-0646 479-0645 479-0656 479-0664 479-1311 479-1310 479-1312 479-1313 479-1314 479-1309 479-1307 479-1308 479-1713 479-1714 479-1105 479-1097 479-1099 479-1109 479-1111 479-1092 479-1090 479-1091 479-1093 479-1094 479-1095 479-1096 479-1098 479-1100 479-1101 479-1102 479-1103 479-1104 479-1106 479-1107 479-1108 479-1112
Racks for chest freezers, Classic
Racks Cryogenic Box Racks
Stainless steel.
  • Racks for different box heights are available
  • Cryoboxes are secured with a locking rod
  • For use with all chest freezer models
The racks offer a storage system that will fit into any chest freezer and can be used with all standard cryoboxes.
Ordering information: Racks are available without cryoboxes or including standard white cardboard boxes.