Cooling thermostats, ECO
Supplier: LAUDA
L001263 L001270 L001277 L001284 L001291 L001298 L001305 L001312 L001319 L001326 L001255
461-1254EA 5440 EUR
461-1254 461-1255 LAUDL001277 LAUDL001284 LAUDL001291 LAUDL001298 LAUDL001305 LAUDL001312 LAUDL001319 LAUDL001326 LAUDL001255
Cooling thermostats, ECO
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Cooling thermostats with menu navigation in plain text for user friendly operation. Various interface modules are available as accessories. All models are available as air or water-cooled variants.
  • Variopump with six adjustable stages
  • Flow rate switch for control of flow rate between internal and external circulation
  • Simultaneous display of set and actual temperature
  • Over-temperature setting via menu
  • USB interface as standard
Silver controller: The cooling thermostats with Silver control head are suitable for use in the temperature range from −50 up to +200 °C. They are equipped with a bath cover and a pump circulation set for external applications as standard.
Heating power 2 kW, working temperature range up to 200 °C
LCD display, resolution of indication 0,01 °C
Programmer with one program and 20 segments
Gold controller: The cooling thermostats with Gold control head are suitable for use in the temperature range from −50 up to +200 °C.
Heating power 2,6 kW, working temperature range up to 200 °C
Colour TFT display, resolution of indication 0,01 °C
Programmer with five programs and 150 segments
Graphical display of temperature profiles
Ramp function and clock timer
Maximum pump capacity:
All models: 22 L/min, 0,55 bar
Delivery information: Units supplied with bath cover and closing plugs. S models have pump connections with 13 mm plastic nipples, G models have a stainless steel pump circulation set with a M16×1 thread. W denotes models are water cooled.
Caution: Cooling capacity at +20; 0; and −20 °C (−15 °C for models RE 415 S and RE 415 G).