Hand disinfectants, Sterillium® Classic Pure
Supplier: Bode
975125 975121 9751220 980390
115-1307EA 52.02 EUR
115-1307 115-4027 115-3993 115-3082
Hand disinfectants, Sterillium® Classic Pure
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
Sterillium® classic pure is particularly suitable for users with sensitive skin. The propanol based hand disinfectant is ideal for use in all areas of the health industry (eg. operating theatres) and hygiene sensitive industries.
  • Comprehensively active against bacteria, fungi and enveloped viruses
  • Easy to use, rub-in product with surgical standard disinfection in 90 seconds
  • Kind to skin with excellent skin tolerance, even with long term use
  • Free from colourants and fragrances
Caution: Use biocides safely. Always read the label and product information before use.
These articles are only available in Germany.