Sodium metatungstate monohydrate ≥85% WO₃ basis, purum p.a., Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Danger
Synonyms: Sodium polytungstate monohydrate, Sodium tungstate (meta) monohydrate, hexa-Sodium tungstate (W12) monohydrate
71913-100G 71913-25G
71913-100GEA 282 EUR
71913-100G 71913-25G
Sodium metatungstate monohydrate ≥85% WO₃ basis, purum p.a., Sigma-Aldrich®
Sodium metatungstate monohydrate
Reagent for the preparation of heavy liquid, for sink-float analysis.
A new medium for density-gradient centrifugation, the density can be adjusted to up to 3.1 g/ml
Formula: Na₆O₃₉W₁₂·H₂O
MW: 2986.13 g/mol (2968.12 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00061466
CAS Number: 12141-67-2
EINECS: 412-770-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eb95f60df8a259a449623cad10a759e96ad60b6&vid=f6c50d2504b787beb1b5804aea58661c1a5498a9&prid=web&clid=SAPDAM