N,N-Dimethylsulphamide

Supplier: Thermo Fisher Scientific
Warning

464680010 464680050
ACRO464680010EA 73.6 EUR
ACRO464680010 ACRO464680050
N,N-Dimethylsulphamide
N,N-Dimethylsulphamide

  • Bioactive

Formula: C₂H₈N₂O₂S
Melting Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD01861286
CAS Number: 3984-14-3

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 95 to 99 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR