Y6, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Fisher Scientific
Synonyms: 2,2'-[[12,13-Bis(2-ethylhexyl)-12,13-dihydro-3,9-diundecylbisthieno[2'',3'':4',5']thieno[2',3':4,5]pyrrolo[3,2-e:2',3'-g][2,1,3]benzothiadiazole-2,10-diyl]bis[methylidyne(5,6-difluoro-3-oxo-1H-indene-2,1(3H)-diylidene)]]bis[propanedinitrile], 2,2'-(((12,13-Bis(2-ethylhexyl)-3,9-diundecyl-12,13-dihydro-[1,2,5]thiadiazolo[3,4-e]thieno[2'',3'':4',5']thieno[2',3':4,5]pyrrolo[3,2-g]thieno[2',3':4,5]thieno[3,2-b]indole-2,10-diyl)bis(methaneylylidene))bis(5,6-difluoro-3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile
H66585.MC H66585.MD H66585.ME
H66585.MCEA 754 EUR
H66585.MC H66585.MD H66585.ME
Y6, Organic Photovoltaics (OPV) reagent
Y6
Formula: C₈₂H₈₆F₄N₈O₂S₅
MW: 1451.93 g/mol
CAS Number: 2304444-49-1
Order Now
Specification Test Results
Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,37 ± 0,02 eV