1,6:2,3-Dianhydro-4-O-(methyl 2,3-di-O-benzyl-β-D-glucopyranosyluronate)-β-D-mannopyranose ≥99%
Supplier: Apollo Scientific
Synonyms: 1,6:2,3-Dianhydro-4-O-(2,3-di-O-benzyl-β-D-glucuronic acid methyl ester)-β-D-mannopyranose
BICL4081-100MG BICL4081-25MG BICL4081-50MG
APOSBICL4081-100MGEA 1490 EUR
APOSBICL4081-100MG APOSBICL4081-25MG APOSBICL4081-50MG
1,6:2,3-Dianhydro-4-O-(methyl 2,3-di-O-benzyl-β-D-glucopyranosyluronate)-β-D-mannopyranose ≥99%
1,6:2,3-Dianhydro-4-O-(methyl 2,3-di-O-benzyl-β-D-glucopyranosyluronate)-β-D-mannopyranose
Storage Temperature: Ambient
CAS Number: 87907-24-2