6-Azido-6-deoxy-D-glucopyranose ≥98%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Azido-6-deoxy-D-glucose
BICL4280-100MG BICL4280-250MG BICL4280-50MG
APOSBICL4280-100MGEA 480 EUR
APOSBICL4280-100MG APOSBICL4280-250MG APOSBICL4280-50MG
6-Azido-6-deoxy-D-glucopyranose ≥98%
6-Azido-6-deoxy-D-glucopyranose
Formula: C₆H₁₁N₃O₅
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD03265537
CAS Number: 20847-05-6