2,3-dimethyl-1-(2,2,2-trichloroethoxysulphuryl)imidazolium triflate
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-dimethyl-1-((2,2,2-trichloroethoxy)sulphonyl)-1H-imidazol-3-ium trifluoromethanesulphonate
BICR143-1G BICR143-5G
APOSBICR143-1GEA 313 EUR
APOSBICR143-1G APOSBICR143-5G
2,3-dimethyl-1-(2,2,2-trichloroethoxysulphuryl)imidazolium triflate
2,3-dimethyl-1-(2,2,2-trichloroethoxysulphuryl)imidazolium triflate
Formula: C₇H₁₀Cl₃N₂O₃S·CF₃O₃S
MW: 457.659 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27664964
CAS Number: 1185733-70-3