Altenuene

Supplier: Apollo Scientific

BIZL0115-1MG BIZL0115-100UG
APOSBIZL0115-1MGEA 696 EUR
APOSBIZL0115-1MG APOSBIZL0115-100UG
Altenuene
Altenuene
Formula: C₁₅H₁₆O₆
Storage Temperature: Ambient
MDL Number: MFCD28143334
CAS Number: 889101-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR