2-Ethynylbenzaldehyde 97%
Supplier: Apollo Scientific
Warning
OR01514-1G OR01514-5G
APOSOR01514-1GEA 34.8 EUR
APOSOR01514-1G APOSOR01514-5G
2-Ethynylbenzaldehyde 97%
2-Ethynylbenzaldehyde
Formula: C₉H₆O
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD04039985
CAS Number: 38846-64-9