2,3-Diphenylpiperazine, 1-BOC protected
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-Diphenylpiperazine-1-carboxylic acid tert-butyl ester
OR01672-100MG OR01672-250MG OR01672-50MG
APOSOR01672-100MGEA 174 EUR
APOSOR01672-100MG APOSOR01672-250MG APOSOR01672-50MG
2,3-Diphenylpiperazine, 1-BOC protected
2,3-Diphenylpiperazine, 1-BOC protected
Formula: C₂₁H₂₆N₂O₂
MW: 338.443 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660371
CAS Number: 886780-65-0