(Morpholin-4-yl)(piperazin-1-yl)methanone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-[(Morpholin-4-yl)carbonyl]piperazine, 4-[(Piperazin-1-yl)carbonyl]morpholine
OR0241-1G OR0241-250MG
APOSOR0241-1GEA 72.5 EUR
APOSOR0241-1G APOSOR0241-250MG
(Morpholin-4-yl)(piperazin-1-yl)methanone 98%
(Morpholin-4-yl)(piperazin-1-yl)methanone
Formula: C₉H₁₇N₃O₂
MW: 199.25 g/mol
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD03844725
CAS Number: 98834-08-3