Ethyl 4-(diethoxyphosphoryl)-3-methylcrotonate, tech
Supplier: Apollo Scientific
Warning
Synonyms: Diethyl [3-(ethoxycarbonyl)-2-methylprop-2-en-1-yl]phosphonate, Ethyl 4-(diethoxyphosphoryl)-3-methylbut-2-enoate
OR10804-5G OR10804-1G
APOSOR10804-5GEA 351 EUR
APOSOR10804-5G APOSOR10804-1G
Ethyl 4-(diethoxyphosphoryl)-3-methylcrotonate, tech
Ethyl 4-(diethoxyphosphoryl)-3-methylcrotonate
Formula: C₁₁H₂₁O₅P
MW: 264.255 g/mol
Boiling Pt: 284…287 °C
Density: 1.089 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD08445507
CAS Number: 41891-54-7