Methyl 4-(5-oxo-1,5-dihydro-4H-1,2,4-triazol-4-yl)benzoate
Supplier: Apollo Scientific
OR110424-1G OR110424-250MG OR110424-5G
APOSOR110424-1GEA 231 EUR
APOSOR110424-1G APOSOR110424-250MG APOSOR110424-5G
Methyl 4-(5-oxo-1,5-dihydro-4H-1,2,4-triazol-4-yl)benzoate
Methyl 4-(5-oxo-1,5-dihydro-4H-1,2,4-triazol-4-yl)benzoate
Formula: C₁₀H₉N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD18384919
CAS Number: 1355171-73-1