4-[4-(tert-Butyl)phenoxy]benzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4-(tert-Butyl)-4'-formyldiphenyl ether, 4-(4-(tert-Butyl)phenoxy)benzaldehyde
OR110518-1G OR110518-5G
APOSOR110518-1GEA 248 EUR
APOSOR110518-1G APOSOR110518-5G
4-[4-(tert-Butyl)phenoxy]benzaldehyde
4-[4-(tert-Butyl)phenoxy]benzaldehyde
Formula: C₁₇H₁₈O₂
MW: 254.324 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08446988
CAS Number: 108934-20-9