Ethyl 4-bromo-1-tert-butyl-5-methyl-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Warning
OR111579-1G OR111579-250MG OR111579-5G
APOSOR111579-1GEA 91.5 EUR
APOSOR111579-1G APOSOR111579-250MG APOSOR111579-5G
Ethyl 4-bromo-1-tert-butyl-5-methyl-1H-pyrazole-3-carboxylate
Ethyl 4-bromo-1-tert-butyl-5-methyl-1H-pyrazole-3-carboxylate
Formula: C₁₁H₁₇BrN₂O₂
MW: 289.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066836
CAS Number: 2391987-03-2