2-Bromo-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)ethan-1-one
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Bromoacetyl)-5-methyl-3-phenyl-1,2-oxazole, [4-(Bromoacetyl)-5-methyl-1,2-oxazol-3-yl]benzene, 4-(Bromoacetyl)-5-methyl-3-phenylisoxazole
OR1233-1G OR1233-5G
APOSOR1233-1GEA 58 EUR
APOSOR1233-1G APOSOR1233-5G
2-Bromo-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)ethan-1-one
2-Bromo-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)ethan-1-one
Formula: C₁₂H₁₀BrNO₂
MW: 280.117 g/mol
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00173902
CAS Number: 104777-39-1