5-(3-Hydroxyphenyl)-1H-pyrazole-3-carboxylic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Carboxy-5-(3-hydroxyphenyl)-1H-pyrazole, 3-(3-Carboxy-1H-pyrazol-5-yl)phenol
OR13182-1G OR13182-250MG
APOSOR13182-1GEA 357 EUR
APOSOR13182-1G APOSOR13182-250MG
5-(3-Hydroxyphenyl)-1H-pyrazole-3-carboxylic acid 97%
5-(3-Hydroxyphenyl)-1H-pyrazole-3-carboxylic acid
Formula: C₁₀H₈N₂O₃
MW: 204.182 g/mol
Melting Pt: 256…257 °C
Storage Temperature: Ambient
MDL Number: MFCD05664420
CAS Number: 690631-98-2