4-Methoxybenzyl bromide, Technical Grade
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-4-methoxybenzene, 4-(Bromomethyl)anisole, 4-(Bromomethyl)phenyl methyl ether
OR16607-1G OR16607-5G OR16607-25G
APOSOR16607-1GEA 53.8 EUR
APOSOR16607-1G APOSOR16607-5G APOSOR16607-25G
4-Methoxybenzyl bromide, Technical Grade
4-Methoxybenzyl bromide
Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 91 °C (1 mm Hg)
Density: 1.395 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00800380
CAS Number: 2746-25-0
UN: 3265
ADR: 8 II