1H-Benzimidazole-4-carboxaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4-Formyl-1H-benzimidazole, 1H-Benzo[d]imidazole-4-carbaldehyde
OR17070-1G OR17070-250MG
APOSOR17070-1GEA 261 EUR
APOSOR17070-1G APOSOR17070-250MG
1H-Benzimidazole-4-carboxaldehyde
1H-Benzimidazole-4-carboxaldehyde
Formula: C₈H₆N₂O
MW: 146.146 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10698555
CAS Number: 144876-36-8