4-(Hydroxymethyl)benzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4-Formylbenzyl alcohol, (4-Formylphenyl)methanol
OR18563-1G OR18563-5G
APOSOR18563-1GEA 69.6 EUR
APOSOR18563-1G APOSOR18563-5G
4-(Hydroxymethyl)benzaldehyde
4-(Hydroxymethyl)benzaldehyde
Formula: C₈H₈O₂
Melting Pt: 43…44 °C
Storage Temperature: Ambient
MDL Number: MFCD11870191
CAS Number: 52010-97-6