2,5-Dibromobenzo[b]furan-3(2H)-one
Supplier: Apollo Scientific
Warning
Synonyms: 2,5-Dibromo-1-benzofuran-3(2H)-one, 2,5-Dibromo-2,3-dihydro-3-oxobenzo[b]furan
OR19511-1G OR19511-250MG
APOSOR19511-1GEA 963 EUR
APOSOR19511-1G APOSOR19511-250MG
2,5-Dibromobenzo[b]furan-3(2H)-one
2,5-Dibromobenzo[b]furan-3(2H)-one
Formula: C₈H₄Br₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD28096610
CAS Number: 1823918-46-2