3-Phenoxypropanoyl chloride
Supplier: Apollo Scientific
OR200139-1G OR200139-250MG
APOSOR200139-1GEA 0 EUR
APOSOR200139-1G APOSOR200139-250MG
3-Phenoxypropanoyl chloride
3-Phenoxypropanoyl chloride
Formula: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11169653
CAS Number: 28317-96-6