5-(Hydroxymethyl)-1H-benzimidazole 97%
Supplier: Apollo Scientific
Warning
Synonyms: (1H-Benzimidazol-5-yl)methanol
OR2056-1G OR2056-250MG
APOSOR2056-1GEA 255 EUR
APOSOR2056-1G APOSOR2056-250MG
5-(Hydroxymethyl)-1H-benzimidazole 97%
5-(Hydroxymethyl)-1H-benzimidazole
Formula: C₈H₈N₂O
MW: 148.162 g/mol
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD08690267
CAS Number: 106429-29-2