Ethyl 3-methyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carboxylate hydrochloride
Supplier: Apollo Scientific
OR24490-1G OR24490-5G
APOSOR24490-1GEA 125 EUR
APOSOR24490-1G APOSOR24490-5G
Ethyl 3-methyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carboxylate hydrochloride
Ethyl 3-methyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carboxylate hydrochloride
Formula: C₉H₁₂N₂O₂S·ClH
MW: 248.73 g/mol
Melting Pt: 194…195 °C
Storage Temperature: Ambient
MDL Number: MFCD01720210
CAS Number: 34467-12-4