N4-(4-chloro-3-nitrophenyl)morpholine-4-carboxamide
Supplier: Apollo Scientific
OR25867-10G OR25867-1G OR25867-5G
APOSOR25867-10GEA 608 EUR
APOSOR25867-10G APOSOR25867-1G APOSOR25867-5G
N4-(4-chloro-3-nitrophenyl)morpholine-4-carboxamide
N4-(4-chloro-3-nitrophenyl)morpholine-4-carboxamide
Formula: C₁₁H₁₂ClN₃O₄
Storage Temperature: Ambient
MDL Number: MFCD00126343
CAS Number: 349118-72-5