5-methyl-1-(2-methylphenyl)-1H-pyrazole-4-carboxylic acid
Supplier: Apollo Scientific
OR25967-1G OR25967-5G
APOSOR25967-1GEA 197 EUR
APOSOR25967-1G APOSOR25967-5G
5-methyl-1-(2-methylphenyl)-1H-pyrazole-4-carboxylic acid
5-methyl-1-(2-methylphenyl)-1H-pyrazole-4-carboxylic acid
Formula: C₁₂H₁₂N₂O₂
MW: 216.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407350
CAS Number: 423768-56-3