tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl ((5-bromothiophen-2-yl)methyl)carbamate
OR25979-1G OR25979-5G
APOSOR25979-1GEA 94.4 EUR
APOSOR25979-1G APOSOR25979-5G
tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate
tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate
Formula: C₁₀H₁₄BrNO₂S
MW: 292.193 g/mol
Melting Pt: 48…53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03659715
CAS Number: 215183-27-0