3-Methyl-1-phenyl-1H-pyrazole-5-carboxylic acid
Supplier: Apollo Scientific
Warning
OR2708-1G OR2708-500MG
APOSOR2708-1GEA 302 EUR
APOSOR2708-1G APOSOR2708-500MG
3-Methyl-1-phenyl-1H-pyrazole-5-carboxylic acid
3-Methyl-1-phenyl-1H-pyrazole-5-carboxylic acid
Formula: C₁₁H₁₀N₂O₂
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD01693509
CAS Number: 1136-76-1