6-(Dimethoxymethyl)-2-thionicotinonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 6-(Dimethoxymethyl)-2-sulphanylpyridine-3-carbonitrile
OR29716-1G OR29716-10G OR29716-5G
APOSOR29716-1GEA 227 EUR
APOSOR29716-1G APOSOR29716-10G APOSOR29716-5G
6-(Dimethoxymethyl)-2-thionicotinonitrile
6-(Dimethoxymethyl)-2-thionicotinonitrile
Formula: C₉H₁₀N₂O₂S
MW: 210.253 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00173693
CAS Number: 175277-23-3
UN: 3439
ADR: 6.1 III