tert-Butyl 2-chloro-5-methylpyridin-4-ylcarbamate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl (2-chloro-5-methylpyridin-4-yl)carbamate
OR300401-1G OR300401-250MG
APOSOR300401-1GEA 578 EUR
APOSOR300401-1G APOSOR300401-250MG
tert-Butyl 2-chloro-5-methylpyridin-4-ylcarbamate
tert-Butyl 2-chloro-5-methylpyridin-4-ylcarbamate
Formula: C₁₁H₁₅ClN₂O₂
MW: 242.702 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26936365
CAS Number: 1820717-60-9