1,1-Dimethylguanidine hydrochloride
Supplier: Apollo Scientific
OR300899-1G OR300899-5G OR300899-250MG
APOSOR300899-1GEA 39.3 EUR
APOSOR300899-1G APOSOR300899-5G APOSOR300899-250MG
1,1-Dimethylguanidine hydrochloride
1,1-Dimethylguanidine hydrochloride
Formula: C₃H₉N₃·ClH
MW: 123.585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152443
CAS Number: 22583-29-5