methyl 4-aminobenzoate hydrochloride
Supplier: Apollo Scientific
OR30151-1G OR30151-250MG OR30151-500MG
APOSOR30151-1GEA 165 EUR
APOSOR30151-1G APOSOR30151-250MG APOSOR30151-500MG
methyl 4-aminobenzoate hydrochloride
methyl 4-aminobenzoate hydrochloride
Formula: C₈H₉NO₂·ClH
MW: 187.623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00205145
CAS Number: 63450-84-0