Thieno[2,3-b]pyridine 97%
Supplier: Apollo Scientific
OR30574-1G OR30574-250MG
APOSOR30574-1GEA 104 EUR
APOSOR30574-1G APOSOR30574-250MG
Thieno[2,3-b]pyridine 97%
Thieno[2,3-b]pyridine
Formula: C₇H₅NS
Storage Temperature: Ambient
MDL Number: MFCD01631145
CAS Number: 272-23-1