3-Bromothieno[2,3-c]pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-6-azabenzo[b]thiophene, 3-Bromo-6-aza-1-benzothiophene
OR30581-1G OR30581-100MG
APOSOR30581-1GEA 149 EUR
APOSOR30581-1G APOSOR30581-100MG
3-Bromothieno[2,3-c]pyridine
3-Bromothieno[2,3-c]pyridine
Formula: C₇H₄BrNS
MW: 214.082 g/mol
Melting Pt: 97…98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11052627
CAS Number: 28783-17-7