(2R)-Morpholine-2-carboxylic acid hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: (R)-Morpholine-2-carboxylic acid hydrochloride
OR308155-1G OR308155-250MG OR308155-5G
APOSOR308155-1GEA 87 EUR
APOSOR308155-1G APOSOR308155-250MG APOSOR308155-5G
(2R)-Morpholine-2-carboxylic acid hydrochloride
(2R)-Morpholine-2-carboxylic acid hydrochloride
Formula: C₅H₉NO₃·ClH
MW: 167.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194711
CAS Number: 1273577-14-2