2-(4-Iodophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Supplier: Apollo Scientific
Warning
OR310872-1G OR310872-5G
APOSOR310872-1GEA 37.7 EUR
APOSOR310872-1G APOSOR310872-5G
2-(4-Iodophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
2-(4-Iodophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Formula: C₁₁H₁₄BIO₂
MW: 315.943 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26394871
CAS Number: 5572-94-1