1,3,5-Trimethyl-1,2-dihydropyridin-2-one
Supplier: Apollo Scientific
OR311009-1G OR311009-500MG
APOSOR311009-1GEA 538 EUR
APOSOR311009-1G APOSOR311009-500MG
1,3,5-Trimethyl-1,2-dihydropyridin-2-one
1,3,5-Trimethyl-1,2-dihydropyridin-2-one
Formula: C₈H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD28097869
CAS Number: 98042-58-1