(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Supplier: Apollo Scientific
Warning
OR315003-1G OR315003-5G OR315003-10G
APOSOR315003-1GEA 21.9 EUR
APOSOR315003-1G APOSOR315003-5G APOSOR315003-10G
(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one
(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Formula: C₆H₇NO
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00211275
CAS Number: 130931-83-8