Diphenylphosphinostyrene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Diphenyl(4-vinylphenyl)phosphine
OR315714-10G
OR315714-1G
OR315714-5G
APOSOR315714-10GEA
161
EUR
APOSOR315714-10G
APOSOR315714-1G
APOSOR315714-5G
Diphenylphosphinostyrene
Diphenylphosphinostyrene
Formula:
C₂₀H₁₇P Melting Pt: 75…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038139 CAS Number: 40538-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...